Learn More
Invitrogen™ NOTUM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139770
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ANMVFIPYCS SDVWSGASSK SEKNEYAFMG ALIIQEVVRE LLGRGLSGAK VLLLAGSSAG GTGVLLNVDR VAEQLEKLGY PAIQVRGLAD SGWFL Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 93%.
Genetic studies in Drosophila have revealed a novel secreted antagonist for Wingless (Wg) signaling, referred to as Notum/Wingful (Wf). Wf/Notum encodes a protein of 430 amino acid residues, which is related to plant pectin acetylesterases of the a/beta-hydrolase superfamily. The main target of Notum action appears to be cell surface proteoglycans Dally and/or Dally-like (Dlp). Notum governs the extracellular Wg morphogen distribution via Dally and Dlp. Also, like some vertebrate Wnt antagonists, notum expression is activated by Wg signaling, revealing the existence of a feedback control mechanism in which Wg modifies its own gradient distribution. In vertebrate, proteoglycans related to Dally and Dlp likely play important roles in Wnt signaling which encodes a glypican that is required for Wnt-11 signaling. A single ortholog of Notum exists in the mouse and the human genome, implying that a similar regulatory mechanism may exist in vertebrates.
Specifications
| NOTUM | |
| Polyclonal | |
| Unconjugated | |
| NOTUM | |
| [Wnt protein] O-palmitoleoyl-L-serine hydrolase; 5730593N15Rik; hNOTUM; NOTUM; notum pectinacetylesterase homolog; notum pectinacetylesterase homolog (Drosophila); NOTUM, palmitoleoyl-protein carboxylesterase; OK/SW-CL.30; Palmitoleoyl-protein carboxylesterase NOTUM; protein notum homolog; RGD1307119 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 147111 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P988 | |
| NOTUM | |
| Recombinant protein corresponding to Human NOTUM. Recombinant protein control fragment (Product #RP-108745). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.