missing translation for 'onlineSavingsMsg'
Learn More

Notoginsenoside R1, TRC

Product Code. 30421457
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421457 10 mg 10mg
30413278 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30421457 Supplier Toronto Research Chemicals Supplier No. TRCN88750010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80418-24-2
Molecular Formula C47 H80 O18
Molecular Weight (g/mol) 933.13
Synonym β-D-Glucopyranoside, (3β,6α,12β)-20-(β-D-glucopyranosyloxy)-3,12-dihydroxydammar-24-en-6-yl 2-O-β-D-xylopyranosyl-, Dammarane, β-D-glucopyranoside deriv., (3β,6α,12β)-20-(β-D-Glucopyranosyloxy)-3,12-dihydroxydammar-24-en-6-yl 2-O-β-D-xylopyranosyl-β-D-glucopyranoside, Notoginsenoside R1, Sanchinoside R1, Sanqi glucoside R1
IUPAC Name (2S,3R,4S,5S,6R)-2-[(2S)-2-[(3S,5R,6S,8R,9R,10R,12R,13R,14R,17S)-6-[(2R,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxyoxan-2-yl]oxy-3,12-dihydroxy-4,4,8,10,14-pentamethyl-2,3,5,6,7,9,11,12,13,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]-6-methylhept-5-en-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CC(=CCC[C@](C)(O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H]2CC[C@]3(C)[C@@H]2[C@H](O)C[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5[C@H](C[C@@]34C)O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O[C@@H]7OC[C@@H](O)[C@H](O)[C@H]7O)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C47H80O18/c1-21(2)10-9-13-47(8,65-41-37(59)34(56)32(54)26(18-48)62-41)22-11-15-45(6)30(22)23(50)16-28-44(5)14-12-29(52)43(3,4)39(44)25(17-46(28,45)7)61-42-38(35(57)33(55)27(19-49)63-42)64-40-36(58)31(53)24(51)20-60-40/h10,22-42,48-59H,9,11-20H2,1-8H3/t22-,23+,24+,25-,26+,27+,28+,29-,30-,31-,32+,33+,34-,35-,36+,37+,38+,39-,40-,41-,42+,44+,45+,46+,47-/m0/s1
Formula Weight 932.5345
Chemical Name or Material Notoginsenoside R1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.