missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notoginsenoside Fe, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Notoginsenoside Fe is a natural compound isolated from Panax japlcus var.
Specifications
Specifications
| Chemical Name or Material | Notoginsenoside Fe |
| CAS | 88105-29-7 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C47H80O17 |
| Quantity | 5 mg |
| Synonym | Notoginseng triterpenes Ginsenoside Mb |
| Solubility Information | H2O : 50 mg/mL (54.52 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble) |
| SMILES | C[C@]12[C@@](C)([C@]3([H])C[C@@H](O)[C@]1([H])[C@@]([H])([C@](C)(CC/C=C(C)/C)O[C@@H]4O[C@H](CO[C@@H]5O[C@@H](CO)[C@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)CC2)CC[C@@]([C@]3(C)CC6)([H])C(C)(C)[C@H]6O[C@@]7([H])[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O7 |
| Molecular Weight (g/mol) | 917.13 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction