missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOTCH4 Recombinant Rabbit Monoclonal Antibody (K01_2K71)

Product Code. 30826390
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826390 100 μL
30828849 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826390 Supplier Invitrogen™ Supplier No. MA563966

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NOTCH4 Recombinant Monoclonal Antibody for Western Blot

This gene encodes a member of the Notch family. Members of this Type 1 transmembrane protein family share structural characteristics including an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. Notch family members play a role in a variety of developmental processes by controlling cell fate decisions. This protein is cleaved in the trans-Golgi network, and presented on the cell surface as a heterodimer. This protein functions as a receptor for membrane bound ligands, and may play a role in vascular, renal and hepatic development. This gene may be associated with susceptibility to schizophrenia in a small portion of cases (entrez gene).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOTCH4
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2K71
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene NOTCH4
Gene Accession No. Q99466
Gene Alias DAQB-143M3.4; FLJ16302; hNotch4; Int3; Int-3; MGC74442; N4; neurogenic locus notch homolog protein 4; Notch 4; Notch 4 extracellular truncation; Notch 4 intracellular domain; Notch gene homolog 4; Notch homolog 4; notch receptor 4; NOTCH3; Notch4; putative Notch4 protein; Transforming protein Int-3
Gene Symbols NOTCH4
Host Species Rabbit
Immunogen A synthetic peptide of human NOTCH4.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4855
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.