Learn More
Invitrogen™ NOTCH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587455
Description
Immunogen sequence: SRLHPVPVPA DWMNRMEVNE TQYNEMFGMV LAPAEGTHPG IAPQSRPPEG KHITTPREPL PPIVTFQLIP KGSIAQPAGA PQPQSTCPPA VAGPLPTMYQ IPEMARLPSV AFPTAMMPQQ DGQVAQTILP AYHPFPASVG KYPTPPSQHS YASSNAAERT PSHSGHLQGE HPYLTPSPES PDQWSSSSPH SASDWSDVTT SPTPGGAGGG QRGPGTHMSE PPHNNMQVYA; Positive Samples: 293T, LO2, HeLa; Cellular Location: Cell membrane, Cytoplasm, Nucleus, Single-pass type I membrane protein.
Neurogenic locus notch homolog protein 2 (NOTCH2) is a highly conserved Notch signaling pathway protein. NOTCH2 is a type 1 transmembrane protein with an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. NOTCH2 functions as a receptor for membrane-bound ligands Jagged1, Jagged2 and Delta1 to regulate cell-fate determination. Defects in NOTCH2 are the cause of Alagille syndrome type 2 (ALGS2). Alagille syndrome is an autosomal dominant multisystem disorder defined clinically by hepatic bile duct paucity and cholestasis in association with cardiac, skeletal, and ophthalmologic manifestations. There are characteristic facial features and less frequent clinical involvement of the renal and vascular systems.
Specifications
| NOTCH2 | |
| Polyclonal | |
| Unconjugated | |
| NOTCH2 | |
| AGS2; AI853703; HJCYS; hN2; Motch B; N2; N2ECD; N2ICD; Neurogenic locus notch homolog protein 2; notch 2; Notch 2 extracellular truncation; Notch 2 intracellular domain; notch gene homolog 2; Notch homolog 2; notch receptor 2; Notch2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18129, 4853 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35516, Q04721 | |
| NOTCH2 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.