missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOTCH2 Monoclonal Antibody (16F11), PE, eBioscience™
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ 12-5786-82

 View more versions of this product

Product Code. 15268829

  • £245.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Description: This 16F11 monoclonal antibody reacts with the extracellular domain of human and mouse Notch2, one of four members of the Notch family of receptors. Notch receptors are 300-kDa single-pass transmembrane proteins. While the extracellular domain contains numerous epidermal growth factor-like repeats for ligand binding, the intracellular domain is involved in cell signaling. Upon binding its membrane-bound ligand (either Delta or Jagged), the Notch receptor undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases the Notch intracellular domain (NICD), which subsequently translocates into the nucleus, heterodimerizes with the DNA-binding protein RBP-J, recruits co-activator molecules, and ultimately activates transcription. Notch2 is expressed on thymocytes (i.e., DN1 and DN2 subsets), activated peripheral T cells, and bone marrow. Moreover, expression of Notch2 has been reported to be altered in various cancers and lymphomas. In addition to its role in stem cell hematopoiesis, Notch2 plays a pivotal role in marginal zone B cell development. Notch2 also interacts with phosphorylated CREB1 to induce granzyme B transcription in cytotoxic T cells. Applications Reported: This 16F11 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 16F11 antibody has been tested by flow cytometric analysis of 24-hour anti-CD3-stimulated mouse splenocytes. This can be use...

Neurogenic locus notch homolog protein 2 (NOTCH2) is a highly conserved Notch signaling pathway protein. NOTCH2 is a type 1 transmembrane protein with an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. NOTCH2 functions as a receptor for membrane-bound ligands Jagged1, Jagged2 and Delta1 to regulate cell-fate determination. Defects in NOTCH2 are the cause of Alagille syndrome type 2 (ALGS2). Alagille syndrome is an autosomal dominant multisystem disorder defined clinically by hepatic bile duct paucity and cholestasis in association with cardiac, skeletal, and ophthalmologic manifestations. There are characteristic facial features and less frequent clinical involvement of the renal and vascular systems.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NOTCH2
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
O35516, Q04721
NOTCH2
Affinity chromatography
RUO
18129, 4853
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
16F11
PE
NOTCH2
AGS2; AI853703; HJCYS; hN2; Motch B; N2; N2ECD; N2ICD; Neurogenic locus notch homolog protein 2; notch 2; Notch 2 extracellular truncation; Notch 2 intracellular domain; notch gene homolog 2; Notch homolog 2; notch receptor 2; Notch2
Rat
100 μg
Primary
Human, Mouse
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.