missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notch-2 Antibody (B6) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-62559-0.025MG
This item is not returnable.
View return policy
Description
Notch-2 Monoclonal specifically detects Notch-2 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence.
Specifications
| Notch-2 | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| NOTCH2 | |
| NRR of mouse Notch 2 | |
| 4853 | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence | |
| B6 | |
| ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence | |
| AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2 | |
| Rabbit | |
| 0.025 mg | |
| An engineered ScFv fragment antibody that binds to both mouse mNRR2 and human hNRR2, but not NRR1. | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction