missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notch-1 Antibody (mN1A) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
£159.00 - £405.00
Specifications
| Antigen | Notch-1 |
|---|---|
| Clone | mN1A |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1 - 2 ug/mL, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunoprecipitation 1:10 - 1:100, Immunohistochemistry-Paraffin 1:10 - 1:500, CyTOF-ready, Knockout Validated |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18068835
|
Novus Biologicals
NB100-78486SS |
0.025 mL |
£159.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18701484
|
Novus Biologicals
NB100-78486 |
0.1 mL |
£405.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Notch-1 Monoclonal specifically detects Notch-1 in Human, Mouse, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready, Knockout Validated.Specifications
| Notch-1 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer Stem Cells, Golgi Apparatus Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| Q01705 | |
| 4851 | |
| Mouse Notch1 protein [Uniprot: Q01705] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 272.505 kDa |
| mN1A | |
| Western Blot 1 - 2 ug/mL, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunoprecipitation 1:10 - 1:100, Immunohistochemistry-Paraffin 1:10 - 1:500, CyTOF-ready, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat (Negative) | |
| EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1 | |
| NOTCH1 | |
| IgG1 κ | |
| Protein G purified | |
| Does not cross-react with Notch 2, 3, or 4. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title