missing translation for 'onlineSavingsMsg'
Learn More

3M™ Nose Cup Assembly 7991

Product Code. 15789827
Click to view available options
:
Color: Black
This item is not returnable. View return policy

Product Code. 15789827

Brand: 3M™ 7000088352

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

3M™ Nose Cup Assembly 7991 includes 3M™ Nose Cup Valve Assembly 7882 for the 3M™ Reusable Full Face Mask 7907S.

  • Replace worn or damaged seals
  • Maintains the right level of protection and comfort
  • Designed for 3M™ Reusable Full Mask 7907S
  • Put on quickly and easily
  • Extend the life of the equipment
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Product Type Nose Cup
For Use With (Equipment) Reusable Full Mask 7907S
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.