missing translation for 'onlineSavingsMsg'
Learn More

Noscapine Hydrochloride, TRC

Product Code. 17991595
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17991595 5 g 5g
17922995 1 g 1g
17951595 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17991595 Supplier Toronto Research Chemicals Supplier No. TRCN8820185G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 912-60-7
Molecular Formula C22 H23 N O7 . Cl H
Molecular Weight (g/mol) 449.88
Synonym 1(3H)-Isobenzofuranone, 6,7-dimethoxy-3-[(5R)-5,6,7,8-tetrahydro-4-methoxy-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-yl]-, hydrochloride (1:1), (3S)-, 1(3H)-Isobenzofuranone, 6,7-dimethoxy-3-(5,6,7,8-tetrahydro-4-methoxy-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-yl)-, hydrochloride, [S-(R*,S*)]-, 1(3H)-Isobenzofuranone, 6,7-dimethoxy-3-[(5R)-5,6,7,8-tetrahydro-4-methoxy-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-yl]-, hydrochloride, (3S)- (9CI), Narcotine, hydrochloride (7CI,8CI), 1,3-Dioxolo[4,5-g]isoquinoline, 1(3H)-isobenzofuranone deriv., Noscapine hydrochloride, Opianine hydrochloride, l-Narcotine hydrochloride
IUPAC Name (3S)-6,7-dimethoxy-3-[(5R)-4-methoxy-6-methyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-5-yl]-3H-2-benzofuran-1-one;hydrochloride
SMILES Cl.COc1ccc2[C@H](OC(=O)c2c1OC)[C@@H]3N(C)CCc4cc5OCOc5c(OC)c34

Specifications

Quantity 5 g
InChI Formula InChI=1S/C22H23NO7.ClH/c1-23-8-7-11-9-14-20(29-10-28-14)21(27-4)15(11)17(23)18-12-5-6-13(25-2)19(26-3)16(12)22(24)30-18;/h5-6,9,17-18H,7-8,10H2,1-4H3;1H/t17-,18+;/m1./s1
Formula Weight 449.1241
Chemical Name or Material Noscapine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.