missing translation for 'onlineSavingsMsg'
Learn More

Norvancomycin Trifluoroacetic Acid Salt >85%, TRC

Product Code. 30423635
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423635 10 mg 10mg
30411996 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30423635 Supplier Toronto Research Chemicals Supplier No. TRCN82531510MG
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 1435.23
Quantity 10 mg
Shipping Condition Room Temperature
Synonym 56-Demethylvancomycin Trifluoroacetic Acid Salt∣Antibiotic A 51568A Trifluoroacetic Acid Salt∣Desmethylvancomycin Trifluoroacetic Acid Salt∣N-Demethylvancomycin Trifluoroacetic Acid Salt∣22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine Vancomycin Derivative Trifluoroacetic Acid Salt∣Demethylvancomycin Trifluoroacetic Acid Salt∣Vancomycin EP Impurity A Trifluoroacetic Acid Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.