Learn More
Nortropinone hydrochloride, 97%, Thermo Scientific Chemicals
£23.30 - £223.00
Chemical Identifiers
CAS | 25602-68-0 |
---|---|
Molecular Formula | C7H12ClNO |
Molecular Weight (g/mol) | 161.629 |
MDL Number | MFCD03613582 |
InChI Key | MZQWQFWRSDNBPV-UHFFFAOYSA-N |
Synonym | nortropinone hydrochloride, 8-azabicyclo 3.2.1 octan-3-one hydrochloride, nortropinone hcl, 8-azabicyclo 3.2.1 octan-3-one hcl, 8-azabicyclo 3.2.1 octan-3-one, hydrochloride, 3-nortropanone hydrochloride, 8-aza-3-oxobicyclo 3.2.1 octane hydrochloride, nortropinonehydrochloride, c7h11no.clh, 8-azabicyclo 3,2,1 octan-3-one hydrochloride |
PubChem CID | 13091218 |
IUPAC Name | 8-azabicyclo[3.2.1]octan-3-one;hydrochloride |
SMILES | C1CC2CC(=O)CC1N2.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15411719
|
Thermo Scientific Alfa Aesar
H64485.06 |
5 g |
£23.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15421719
|
Thermo Scientific Alfa Aesar
H64485.14 |
25 g |
£69.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15431719
|
Thermo Scientific Alfa Aesar
H64485.22 |
100 g |
£223.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25602-68-0 | |
161.629 | |
MZQWQFWRSDNBPV-UHFFFAOYSA-N | |
13091218 | |
C1CC2CC(=O)CC1N2.Cl |
C7H12ClNO | |
MFCD03613582 | |
nortropinone hydrochloride, 8-azabicyclo 3.2.1 octan-3-one hydrochloride, nortropinone hcl, 8-azabicyclo 3.2.1 octan-3-one hcl, 8-azabicyclo 3.2.1 octan-3-one, hydrochloride, 3-nortropanone hydrochloride, 8-aza-3-oxobicyclo 3.2.1 octane hydrochloride, nortropinonehydrochloride, c7h11no.clh, 8-azabicyclo 3,2,1 octan-3-one hydrochloride | |
8-azabicyclo[3.2.1]octan-3-one;hydrochloride |
Specifications
25602-68-0 | |
C7H12ClNO | |
3693524 | |
nortropinone hydrochloride, 8-azabicyclo 3.2.1 octan-3-one hydrochloride, nortropinone hcl, 8-azabicyclo 3.2.1 octan-3-one hcl, 8-azabicyclo 3.2.1 octan-3-one, hydrochloride, 3-nortropanone hydrochloride, 8-aza-3-oxobicyclo 3.2.1 octane hydrochloride, nortropinonehydrochloride, c7h11no.clh, 8-azabicyclo 3,2,1 octan-3-one hydrochloride | |
C1CC2CC(=O)CC1N2.Cl | |
161.629 | |
161.63 | |
Solid |
194°C to 196°C | |
MFCD03613582 | |
Hygroscopic | |
MZQWQFWRSDNBPV-UHFFFAOYSA-N | |
8-azabicyclo[3.2.1]octan-3-one;hydrochloride | |
13091218 | |
≥98% | |
Nortropinone hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only