missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nortriptyline Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA175481
This item is not returnable.
View return policy
Description
Working Concentration: 5 μg/mL. Sensitivity: 10 ng/mL Nortriptyline produces 45% inhibition in a competitive ELISA employing Tricyclic Antidepressants (TCA) (4) polyclonal antibody.
Nortriptyline Base is a tricyclic antidepressant agent used for short-term treatment of various forms of depression. Nortriptyline blocks the norepinephrine presynaptic receptors, thereby blocking the reuptake of this neurotransmitter and raising the concentration in the synaptic cleft in the CNS. Nortriptyline also binds to alpha-adrenergic, histaminergic and cholinergic receptors. Long-term treatment with nortriptyline produces a downregulation of adrenergic receptors due to the increased stimulation of these receptors. [National Center for Biotechnology Information. PubChem Compound Database; CID=4543].
Specifications
| Nortriptyline | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.1 | |
| Sheep | |
| Purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA | |
| Unconjugated | |
| Aventyl; C19H21N; Desitriptilina; Desmethylamitriptyline; Noritren; Sensaval | |
| Nortriptyline-BTG. | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction