missing translation for 'onlineSavingsMsg'
Learn More

Nortriptyline N-Ethyl Carbamate, TRC

Product Code. 30425894
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425894 10 g 10g
30417298 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30425894 Supplier Toronto Research Chemicals Supplier No. TRCN83690010G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16234-88-1
Molecular Formula C22 H25 N O2
Molecular Weight (g/mol) 335.44
Synonym Ethyl [3-(10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-ylidene)propyl]methylcarbamate, Nortriptyline N-Ethyl Carbamate
IUPAC Name ethyl N-[3-(5,6-dihydrodibenzo[1,2-a:2',1'-d][7]annulen-11-ylidene)propyl]-N-methylcarbamate
SMILES CCOC(=O)N(C)CCC=C1c2ccccc2CCc3ccccc13

Specifications

Quantity 10 g
InChI Formula InChI=1S/C22H25NO2/c1-3-25-22(24)23(2)16-8-13-21-19-11-6-4-9-17(19)14-15-18-10-5-7-12-20(18)21/h4-7,9-13H,3,8,14-16H2,1-2H3
Formula Weight 335.1885
Chemical Name or Material Ethyl [3-(10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-ylidene)propyl]methylcarbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.