missing translation for 'onlineSavingsMsg'
Learn More

Nortriptyline-D3 Hydrochloride 0.1 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30463679
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463679 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30463679 Supplier LoGiCal Supplier No. LGCAMP0079.8002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nortriptyline-D3 Hydrochloride
CAS 203784-52-5
Solvent or Matrix Methanol
Molecular Formula C19 2H3 H18 N . Cl H
Quantity 1 mL
Synonym Nortriptyline-D3 Hydrochloride, 3-(10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-(methyl-d3)-1-propanamine hydrochloride, 1-Propanamine, 3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-(methyl-d3)-, hydrochloride (1:1), 1-Propanamine, 3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-(methyl-d3)-, hydrochloride (9CI), 3-(10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-(methyl-d3)-1-propanamine hydrochloride (1:1)
InChI Formula InChI=1S/C19H21N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-11,20H,6,12-14H2,1H3;1H/i1D3;
SMILES Cl.[2H]C([2H])([2H])NCCC=C1c2ccccc2CCc3ccccc13
IUPAC Name 3-(5,6-dihydrodibenzo[1,2-a:2',1'-d][7]annulen-11-ylidene)-N-(trideuteriomethyl)propan-1-amine;hydrochloride
Molecular Weight (g/mol) 302.86
Formula Weight 302.1629 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.