Learn More
Thermo Scientific™ North2South™ Complete Biotin Random Prime Labeling and Detection Kit
Acquire consistent, highly sensitive Southern blot results with a novel ECL HRP substrate and optimized hybridization and blocking buffers.
Brand: Thermo Scientific™ 17175
Code : PN
Additional Details : Weight : 0.25000kg
Description
Thermo Scientific™ North2South Chemiluminescent Detection Kit combines a novel ECL substrate for horseradish peroxidase (HRP) and optimized hybridization and blocking buffers to ensure consistent, high-sensitivity Southern blot results.
The Thermo Scientific North2South Hybridization and Detection Kit combines a novel enhanced luminol substrate for horseradish peroxidase (HRP) with optimized pre-made hybridization and blocking buffers to ensure consistent results and high sensitivity. Any nucleic acid blot can be hybridized with the appropriate biotinylated (biotin-labeled) oligonucleotide probe. After a brief membrane blocking step, the biotin label is detected with streptavidin-HRP and a 5-minute incubation with the chemiluminescent substrate. Results are easily recorded by brief exposure to X-ray film (1-15 minutes) or CCD camera.Highlights:
- Sensitivity equal to or greater than 32P
- Amenable to many formats
- High level of light output and consistent low background allows for very short exposure times using cooled CCD camera systems or film
- Ready-to-use buffers and short process time makes switching from radioactive detection to chemiluminescent detection an easy transition, even for the novice user
Specifications
Sufficient For: 10 biotin-labeling reactions (each with 100 ng DNA) and 10 mini-gel blots • North2South Chemiluminescent Hybridization and Detection Kit (Part No. 17097), 1 kit • North2South Biotin Random Prime Labeling Kit (Part No. 17075), 1 kit |
|
Kit | |
Biotin Labeling and Detection Kit |
Chromogenic | |
North2South™ | |
2164 mL Kit |