missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ North2South™ Biotin Random Prime DNA Labeling Kit

Product Code. 10229423 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 Reactions
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10229423

Brand: Thermo Scientific™ 17075

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Produce biotinylated DNA templates for DNA hybridization and detection methods using random heptanucleotides, Klenow fragment and biotin-nucleotides.

Thermo Scientific™ North2South Random Prime DNA Biotinylation Kit uses random heptanucleotides, Klenow fragment and biotin-nucleotides to produce biotinylated DNA templates for DNA hybridization and detection methods.

Random heptanucleotides containing all possible sequences are annealed to a denatured DNA template. These act as primers for complementary strand synthesis by DNA polymerase. Biotinylated dNTPs in the reaction mix are incorporated into the newly synthesized DNA. This protocol yields biotin-labeled DNA of high activity for use as probes in hybridization experiments such as Southern and Northern hybridizations. The resulting hybrids can be detected with streptavidin-horseradish peroxidase (HRP).

Highlights:

  • Ultimate sensitivity - labels with biotinylated dUTP, maximizing streptavidin-HRP binding and increasing sensitivity
  • Works with as little as 100ng starting DNA template
  • Exonuclease-free Klenow fragment produces higher yields
  • Each reaction yields probe sufficient for approximately three (10 x 10cm) blots
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sufficient For: 10 biotin-labeling reactions (each with 100 ng DNA)
• Heptanucleotide Mix (5X), 100 μL
• Deoxynucleotide Mix (5X), 80 μL
• Reaction Buffer (10X), 50 μL
• DNA Polymerase Klenow Fragment (5U/μL), 10 μL
• Control DNA (250 ng/μL), 5 μL
• Ammonium Acetate (5M), 1 mL
• EDTA (0.5M, pH 8.0), 1 mL
• Nuclease-free Water, 1 mL
• Biotin-11-dUTP, 20 μL

Store all components at -20°C.
Format Kit
Labeling Target DNA
Product Line North2South
Product Type DNA Labeling Kit
Quantity 10 Reactions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.