missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nortestosterone Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA175005
This item is not returnable.
View return policy
Description
Working Concentration: 0.25 μg/mL. Sensitivity: 10 ng/mL 17alpha-19 Nortestosterone produces 93% inhibition in a competitive ELISA employing 17α-Nortestosterone polyclonal antibody.
Nandrolone is a synthetic, anabolic steroid analog of testosterone. Similar to testosterone, nortestosterone (19-Nortestosterone) binds to and activates specific nuclear receptors. This agent may be used for testosterone replacement therapy in hypogonadal men, in HIV-wasting syndrome, and in other conditions in order to increase nitrogen retention and fat-free muscle mass. [National Center for Biotechnology Information. PubChem Compound Database; CID=9904].
Specifications
| Nortestosterone | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Sheep | |
| Purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA | |
| Unconjugated | |
| 19-Norandrostenolone; 19-Nortestosterone; C18H26O2; Nandrolone; Norandrostenolone; Nortestosterone | |
| 17a-19-Nortestosterone(3)-BTG. | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction