missing translation for 'onlineSavingsMsg'
Learn More

Nortadalafil, MedChemExpress

Product Code. 16498248
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498248 10 mg 10mg
16470698 5 mg 5mg
16464137 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498248 Supplier Medchem Express Supplier No. HY90009
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nortadalafil is demethyl Tadalafil, which is a PDE5 inhibitor, currently marketed in pill form for treating erectile dysfunction (ED) under the name Cialis; and under the name Adcirca for the treatment of pulmonary arterial hypertension.

Nortadalafil is demethyl Tadalafil, which is a PDE5 inhibitor, currently marketed in pill form for treating erectile dysfunction (ED) under the name Cialis

Specifications

Chemical Name or Material Nortadalafil
CAS 171596-36-4
Color White
Purity Grade Notes Research
Molecular Formula C21H17N3O4
Quantity 10 mg
Synonym Demethyl Tadalafil
Solubility Information DMSO : 50 mg/mL (133.20 mM; Need ultrasonic)
SMILES O=C1NCC(N([C@@H]1C2)[C@H](C3=CC=C(OCO4)C4=C3)C5=C2C6=C(N5)C=CC=C6)=O
Molecular Weight (g/mol) 375.38
Formula Weight 375.38
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.