missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norovirus VP1 Antibody (A1431) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43557
This item is not returnable.
View return policy
Description
Norovirus VP1 Monoclonal antibody specifically detects Norovirus VP1 in Virus samples. It is validated for ELISA,Neutralization
Specifications
| Norovirus VP1 | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA, Block/Neutralize | |
| Capsid Protein, Capsid protein VP1, CP, Norovirus Capsid protein VP1, Norwalk virus VP1, NoV VP1, p59 | |
| The original antibody was isolated from an anti-NoV GII.4c IgG serum antibody repertoire generated from three donors that experienced a significant increase in GII.4 titer after immunization with the bivalent GII.4c + GI.1 VLP vaccine. (Uniprot# S5YAA0) | |
| 0.2 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Neutralization | |
| A1431 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction