missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Normal Porcine Serum

Product Code. 11839230
Change view
Click to view available options
Quantity:
120 mg
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11839230 120 mg 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11839230 Supplier Invitrogen™ Supplier No. 31890
  • ONLINE EXCLUSIVE PRICE
    £64.25
    £48.65 / 2mL
    Save £15.60
    24% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Porcine Control

For blocking purposes, use 5% (v/v) solution (1:20 dilution from rehydrated volume). Rehydration according to instruction yields 100% serum. Restoration and Storage: Store product at 4°C until opened. Restore with 2.0 mL distilled water. Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, freeze rehydrated undiluted product in small aliquots at 20°C. Country of Origin: USA.

Normal Sera are non-immune serum samples from goat, human, mouse, rabbit and other species that contain the usual complement of serum proteins, including the various classes of immunoglobulins that exist in healthy animals of the particular species. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components. Normal serum is frequently used for blocking or saturating generalized binding interactions for immunodetection methods, especially those involving tissue samples such as immunohistochemistry (IHC). Normal sera are also useful as controls for testing general and specific antibody purification methods.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Form Lyophilized
Product Type Normal Porcine Serum
Quantity 120 mg
Concentration 60.0 mg/mL
Formulation Whole serum, PBS with 0.05% sodium azide; pH 7.6
For Use With (Application) Immunohistochemistry, Immunocytochemistry, Control, Blocking Assay
Isotype Ig
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.