Learn More
GPE Scientific™ NorLoc™ NMR Tube Caps
Product Code. 15858254
Change view
Click to view available options
Color:
Black Onyx
Blue Royal
Glacier White
Green Envy
Lemon Yellow
Red Poppy
Unit Size:
Pack of 100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15858254 Blue Royal -
15638995 Black Onyx -
15648995 Glacier White Pack of 100
15658995 Lemon Yellow -
15668995 Red Poppy Pack of 100
15868254 Green Envy -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15858254 Supplier GPE Scientific™ Supplier No. NORLOC25BR100PK
Only null left
Add to basket
Add to basket
GPE Scientific™ NorLoc™ NMR Tube Caps can be applied easily and quickly, increasing personal safety and saving valuable time.
  • Features a patented design that addresses many of the flaws in the traditional NMR tube cap.
  • Can be applied much more easily and quickly, thereby increasing personal safety and saving valuable time.
  • Incorporates an advanced seal design, conferring superior holding and sealing capabilities.
  • Superior retention of the cap to the tube forms a vaulted seal that safeguards precious or critical samples, even during refrigerated cold storage and variable temperature cycling and withstands repeated removals and reapplications
Specifications
Product Type NMR Tube Cap
Color Blue Royal
Quantity 100/Pk.
Size 5 mm