missing translation for 'onlineSavingsMsg'
Learn More

Norlaudanosoline Hydrobromide, TRC

Product Code. 30136990
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136990 10 mg 10mg
30138225 100 mg 100mg
30009410 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136990 Supplier Toronto Research Chemicals Supplier No. TRCN66142510MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16659-88-4
Molecular Formula C16 H17 N O4 . Br H
Molecular Weight (g/mol) 368.22
Synonym 1-[(3,4-Dihydroxyphenyl)methyl]-1,2,3,4-tetrahydro-6,7-isoquinolinediol Hydrobromide (1:1), 1-(3,4-Dihydroxybenzyl)-1,2,3,4-tetrahydro-6,7-isoquinolinediol Hydrobromide, 1(±)-Tetrahydropapaveroline Hydrobromide, NSC 118071, Tetrahydropapaveroline Bromide, Tetrahydropapaveroline Hydrobromide
IUPAC Name 1-[(3,4-dihydroxyphenyl)methyl]-1,2,3,4-tetrahydroisoquinoline-6,7-diol;hydrobromide
SMILES Br.Oc1ccc(CC2NCCc3cc(O)c(O)cc23)cc1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H17NO4.BrH/c18-13-2-1-9(6-14(13)19)5-12-11-8-16(21)15(20)7-10(11)3-4-17-12;/h1-2,6-8,12,17-21H,3-5H2;1H
Formula Weight 367.0419
Percent Purity >95%
Chemical Name or Material Norlaudanosoline Hydrobromide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.