missing translation for 'onlineSavingsMsg'
Learn More

Norisoboldine, MedChemExpress

Product Code. 16439028
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16439028 10 mg 10mg
16421448 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16439028 Supplier Medchem Express Supplier No. HYN0586
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Norisoboldine is an orally active natural aryl hydrocarbon receptor (AhR) agonist. Norisoboldine, as a major isoquinoline alkaloid present in Radix Linderae, can be used for the research of Rheumatoid arthritis and Ulcerative colitis.

Norisoboldine is an orally active natural aryl hydrocarbon receptor (AhR) agonist. Norisoboldine, as a major isoquinoline alkaloid present in Radix Linderae, can be used for the research of Rheumatoid arthritis and Ulcerative colitis.

Specifications

Chemical Name or Material Norisoboldine
CAS 23599-69-1
Color Brown
Purity Grade Notes Research
Molecular Formula C18H19NO4
Quantity 10 mg
Synonym (+)-Laurelliptine
Solubility Information DMSO : ≥ 62.5 mg/mL (199.46 mM)
SMILES OC1=C(OC)C=C2CCN[C@@]3([H])CC4=CC(O)=C(OC)C=C4C1=C23
Molecular Weight (g/mol) 313.35
Formula Weight 313.35
Percent Purity 99.44%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.