missing translation for 'onlineSavingsMsg'
Learn More

Norharman, 98%

Product Code. 10598883 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
100mg
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10598883 100 mg Glass bottle 100mg
10021430 500 mg Glass bottle 500mg
10618454 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10598883 Supplier Thermo Scientific Acros Supplier No. 129261000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 244-63-3 | C11H8N2 | 168.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 244-63-3
Molecular Formula C11H8N2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00004956
InChI Key AIFRHYZBTHREPW-UHFFFAOYSA-N
Synonym 9h-pyrido 3,4-b indole, norharman, norharmane, beta-carboline, carbazoline, 2,9-diazafluorene, 9h-beta-carboline, 2-azacarbazole, .beta.-carboline
PubChem CID 64961
ChEBI CHEBI:109895
IUPAC Name 9H-pyrido[3,4-b]indole
SMILES N1C2=C(C=CC=C2)C2=C1C=NC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 199.0°C to 201.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Tertiary base)
Packaging Glass bottle
Quantity 100 mg
Beilstein 23, II, 224
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 168.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Norharman
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.