missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norfluoxetine Oxalate 1.0 mg/ml in Methanol (as free base), LoGiCal
Description
Norfluoxetine Oxalate 1.0 mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Norfluoxetine oxalate |
| CAS | 107674-50-0 |
| Analyte or Component Names | Norfluoxetine oxalate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H16 F3 N O . C2 H2 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Norfluoxetine Oxalate |
| InChI Formula | InChI=1 S/C16H16F3NO.C2H2O4/c17-16(18,19)13-6-8-14(9-7-13)21-15(10-11-20)12-4-2-1-3-5-12;3-1(4)2(5)6/h1-9,15 H,10-11,20H2;(H,3,4)(H,5,6) |
| SMILES | NCCC(Oc1ccc(cc1)C(F)(F)F)c2ccccc2.OC(=O)C(=O)O |
| IUPAC Name | oxalic acid;3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?