missing translation for 'onlineSavingsMsg'
Learn More

Lipomed Norfluoxetine-D6.oxalate

Product Code. 30458063
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458063 10 mg 10mg
30460609 50 mg 50mg
30461409 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30458063 Supplier Lipomed Supplier No. LPMFLX1859OX10
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Lipomed's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Norfluoxetine-D6 oxalate
CAS 1435934-41-0
Molecular Formula C16 D6 H10 F3 N O . C2 H2 O4
Quantity 10 mg
Synonym Norfluoxetine-D6 Oxalate
InChI Formula InChI=1 S/C16H16F3NO.C2H2O4/c17-16(18,19)13-6-8-14(9-7-13)21-15(10-11-20)12-4-2-1-3-5-12;3-1(4)2(5)6/h1-9,15 H,10-11,20H2;(H,3,4)(H,5,6)/i1D,2 D,3 D,4 D,5 D,15 D;
SMILES [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])(CCN)Oc2ccc(cc2)C(F)(F)F.OC(=O)C(=O)O
IUPAC Name 3-deuterio-3-(2,3,4,5,6-pentadeuteriophenyl)-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;oxalic acid
Molecular Weight (g/mol) 391.37
Formula Weight 391.1514
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.