missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norfloxacin, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Norfloxacin
Specifications
Specifications
| Chemical Name or Material | Norfloxacin |
| CAS | 70458-96-7 |
| Analyte or Component Names | Norfloxacin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H18 F N3 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 3-Quinolinecarboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-, 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid, 1-Ethyl-6-fluoro-1,4-dihydro-7-(1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid, 4803P, AM 715, Amicrobin, Baccidal, Barazan, Barotham, Chibroxin, Chibroxine, Chibroxol, Esclebin, Floxacin, Floxacin 400, Fulgram, Gonorcin, Lexinor, Mk 0366, N-Demethylpefloxacin, N-Desmethylpefloxacin, Neofloxacin, Neofloxin, Noflo, Nofocin, Nolicin, Noracin, Noraxin, Norbid, Norflox, Norflox 400, Norfloxacin, Norfloxacin Bayvit, Norfloxacine, Norfloxacine EG, Norfloxacine Ratiopharm, Norfloxacine Teva, Norfloxin, Norilet, Normax, Norocin, Noroxacine, Noroxin, Noroxine, Norphloxacine, Norxacin, Renosept, Sebercim, Spectrama, Urekacin, Uriflox, Uroflox, Urotrobel, Uroxacin, Utin, Utinor, Zoroxin, Pefloxacin Mesilate Dihydrate Impurity A, Pefloxacin Impurity A |
| InChI Formula | InChI=1S/C16H18FN3O3/c1-2-19-9-11(16(22)23)15(21)10-7-12(17)14(8-13(10)19)20-5-3-18-4-6-20/h7-9,18H,2-6H2,1H3,(H,22,23) |
| SMILES | CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCNCC3 |
| IUPAC Name | 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction