missing translation for 'onlineSavingsMsg'
Learn More

Norfloxacin, MedChemExpress

Product Code. 16480778
Encompass_Preferred
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 16480778

Brand: Medchem Express HYB0132

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Norfloxacin (MK-0366) is a broad-spectrum antibiotic that is active against both Gram-positive and Gram-negative bacteria, which functions by inhibiting DNA gyrase.

Norfloxacin (MK-0366) is a broad-spectrum antibiotic that is active against both Gram-positive and Gram-negative bacteria, which functions by inhibiting DNA gyrase.

Specifications

Chemical Name or Material Norfloxacin
Physical Form Solid
CAS 70458-96-7
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C16H18FN3O3
For Use With (Application) COVID-19-immunoregulation
Synonym MK-0366
Solubility Information DMSO : 5 mg/mL (15.66 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CN(CC)C2=C(C=C(F)C(N3CCNCC3)=C2)C1=O)O
Molecular Weight (g/mol) 319.33
Formula Weight 319.33
Percent Purity 95.45%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.