missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norfloxacin Ethyl Ester, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Norfloxacin Ethyl Ester
Specifications
Specifications
| Chemical Name or Material | Norfloxacin Ethyl Ester |
| CAS | 74011-47-5 |
| Analyte or Component Names | Norfloxacin Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H22 F N3 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Norfloxacin Ethyl Ester, 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid ethyl ester, A 56199, Ethyl norfloxacin, Norfloxacin ethyl ester |
| InChI Formula | InChI=1S/C18H22FN3O3/c1-3-21-11-13(18(24)25-4-2)17(23)12-9-14(19)16(10-15(12)21)22-7-5-20-6-8-22/h9-11,20H,3-8H2,1-2H3 |
| SMILES | CCOC(=O)C1=CN(CC)c2cc(N3CCNCC3)c(F)cc2C1=O |
| IUPAC Name | ethyl 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction