missing translation for 'onlineSavingsMsg'
Learn More

Norfloxacin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30318235
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318235 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318235 Supplier Dr. Ehrenstorfer Supplier No. DREA15648000AL100
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Norfloxacin
CAS 70458-96-7
Analyte or Component Names Norfloxacin
Shipping Condition Room Temperature
Molecular Formula C16 H18 F N3 O3
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 3-Quinolinecarboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-, 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid, 1-Ethyl-6-fluoro-1,4-dihydro-7-(1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid, 4803P, AM 715, Amicrobin, Baccidal, Barazan, Barotham, Chibroxin, Chibroxine, Chibroxol, Esclebin, Floxacin, Floxacin 400, Fulgram, Gonorcin, Lexinor, Mk 0366, N-Demethylpefloxacin, N-Desmethylpefloxacin, Neofloxacin, Neofloxin, Noflo, Nofocin, Nolicin, Noracin, Noraxin, Norbid, Norflox, Norflox 400, Norfloxacin, Norfloxacin Bayvit, Norfloxacine, Norfloxacine EG, Norfloxacine Ratiopharm, Norfloxacine Teva, Norfloxin, Norilet, Normax, Norocin, Noroxacine, Noroxin, Noroxine, Norphloxacine, Norxacin, Renosept, Sebercim, Spectrama, Urekacin, Uriflox, Uroflox, Urotrobel, Uroxacin, Utin, Utinor, Zoroxin, Pefloxacin Mesilate Dihydrate Impurity A, Pefloxacin Impurity A
InChI Formula InChI=1S/C16H18FN3O3/c1-2-19-9-11(16(22)23)15(21)10-7-12(17)14(8-13(10)19)20-5-3-18-4-6-20/h7-9,18H,2-6H2,1H3,(H,22,23)
SMILES CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCNCC3
IUPAC Name 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 319.33
Quantity 1 mL
Formula Weight 319.1332 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.