missing translation for 'onlineSavingsMsg'
Learn More

Norethisterone, Thermo Scientific Chemicals

Product Code. 15507039 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15507039

Brand: Thermo Scientific Acros 262890050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-22-4
Molecular Formula C20H26O2
Molecular Weight (g/mol) 298.42
InChI Key VIKNJXKGJWUCNN-XGXHKTLJSA-N
Synonym norethindrone, norethisterone, norethisteron, micronor, 19-norethisterone, norcolut, noriday, norlutin, primolut-n, norethynodrone
PubChem CID 6230
ChEBI CHEBI:7627
IUPAC Name (8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES CC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34

Specifications

Melting Point 205.0°C to 206.0°C
Loss on Drying 0.5% max. (1 g, 105°C, 3 h)
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, dioxane
Formula Weight 298.42
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Norethisterone
compliance-icons compliance-icons
Product Identifier
  • Norethisterone
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity on or via lactation
  • Reproductive toxicity Category 1A
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H362-May cause harm to breast-fed children.
  • H351-Suspected of causing cancer.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P263-Avoid contact during pregnancy/while nursing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.