missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norethisterone Enantate (Norethindrone Enanthate), Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Norethisterone Enantate (Norethindrone Enanthate)
Specifications
Specifications
| Chemical Name or Material | Norethisterone Enantate (Norethindrone Enanthate) |
| CAS | 3836-23-5 |
| Analyte or Component Names | Norethisterone Enantate (Norethindrone Enanthate) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H38 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | (17α)-17-[(1-oxoheptyl)oxy]-19-Norpregn-4-en-20-yn-3-one, 19-Nor-17α-pregn-4-en-20-yn-3-one, 17-hydroxy-, heptanoate (6CI,7CI,8CI), Heptanoic acid, ester with 17-hydroxy-19-nor-17α-pregn-4-en-20-yn-3-one (8CI), 17α-Ethynyl-19-nortestosterone 17-heptanoate, 17α-Ethynyl-19-nortestosterone enanthate, NSC 22846, Norethindrone enanthate, Norethisterone enanthate, Norethisterone heptanoate, Norigest, Noristerat, SH 8.0393, ZK 5410 |
| InChI Formula | InChI=1S/C27H38O3/c1-4-6-7-8-9-25(29)30-27(5-2)17-15-24-23-12-10-19-18-20(28)11-13-21(19)22(23)14-16-26(24,27)3/h2,18,21-24H,4,6-17H2,1,3H3/t21-,22+,23+,24-,26-,27-/m0/s1 |
| SMILES | CCCCCCC(=O)O[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C#C |
| IUPAC Name | [(8R,9S,10R,13S,14S,17R)-17-ethynyl-13-methyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] heptanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction