missing translation for 'onlineSavingsMsg'
Learn More

Norethindrone, TRC

Product Code. 30409828
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30409828 1 g
30421959 250 mg
30422560 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30409828 Supplier Toronto Research Chemicals Supplier No. TRCN6760001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-22-4
Molecular Formula C20 H26 O2
Molecular Weight (g/mol) 298.42
Synonym Norethisterone Acetate Imp. A (EP), Levonorgestrel Imp. U (EP), (17α)-17-Hydroxy-19-Norpregn-4-en-20-yn-3-one, 17-Hydroxy-19-nor-17α-pregn-4-en-20 yn-3-one, 17α-Ethinyl-17β-hydroxy-δ4-estren-3-one, 17α-Ethinyl-19-nortestosterone, 17α-Ethinylestr-4-en-17β-ol-3-one, 17α-Ethynyl-17-hydroxy-4-estrene-3-one, 17α-Ethynyl-17-hydroxyest-4-en-3-one, 17α-Ethynyl-17-hydroxyestr-4-en-3-one, 17α-Ethynyl-17β-hydroxy-19-norandrost-4-en-3-one, 17α-Ethynyl-17β-hydroxyestr-4-en-3-one, 17α-Ethynyl-19-nor-androst-4-en-17β-ol-3-one, 17α-Ethynyl-19-nortestosterone, 17α-Ethynyl-3-oxo-4-estren-17β-ol, 17β-Hydroxy-17α-ethynylestr-4-en-3-one, 19-Nor-17α-ethynyl-17β-hydroxy-4-androsten-3-one, 19-Nor-17α-ethynylandrosten-17β-ol-3-one, 19-Nor-17α-ethynyltestosterone, 19-Norandrost-4-en-3-one, 17α-ethynyl-17β-hydroxy-, 19-Nortestosterone, 17-ethynyl-, Anovule, Conludaf, Conludag, Estr-4-ene-17α-ethynyl-17β-ol-3-one, Ethinylnortestosterone, Ethynylnortestosterone, Gestest, Menzol, Micronett, Micronor, Micronovum, Mini-Pe, Mini-pill, NSC 9564, Nor-QD, Noralutin, Norcolut, Norethindrone, Norethisteron, Norethisterone, Norethynodrone, Noretisterone Dak, Norfor, Norgestin, Norluten, Norlutin, Norluton, Normapause, Norpregneninolone, Primolut N, Proluteasi, SC 4640, Triella, Utovlan
IUPAC Name (8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H26O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,12,15-18,22H,4-11H2,2H3/t15-,16+,17+,18-,19-,20-/m0/s1
Formula Weight 298.1933
Percent Purity >95%
Chemical Name or Material Norethisterone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.