missing translation for 'onlineSavingsMsg'
Learn More

Norethindrone, MP Biomedicals™

Product Code. 11499231 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499231 5 g 5g
11479231 0.25 g 250mg
11489231 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499231 Supplier MP Biomedicals Supplier No. 0215594694

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 68-22-4
Molecular Formula C20H26O2
Molecular Weight (g/mol) 298.426
InChI Key VIKNJXKGJWUCNN-XGXHKTLJSA-N
Synonym norethindrone, norethisterone, norethisteron, micronor, 19-norethisterone, norcolut, noriday, norlutin, primolut-n, norethynodrone
PubChem CID 6230
ChEBI CHEBI:7627
IUPAC Name (8R,9S,10R,13S,14S,17R)-17-ethynyl-17-hydroxy-13-methyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES CC12CCC3C(C1CCC2(C#C)O)CCC4=CC(=O)CCC34

Spécification

Color White
Quantity 5 g
Formula Weight 298.4
Physical Form Powder
Chemical Name or Material Norethindrone
compliance-icons
Identifiant produit
  • Norethindrone
Mention d'avertissement
  • Danger
Catégorie de danger
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1A
Mention de danger
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H362-May cause harm to breast-fed children.
Conseils de prudence
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P263-Avoid contact during pregnancy/while nursing.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Informations supplémentaires
  • MIXTURE LIST-Contains : norethindrone
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.