missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norethindrone, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
1g
250mg
5g
Specifications
Specifications
| Color | White |
| Quantity | 0.25 g |
| Formula Weight | 298.4 |
| Physical Form | Powder |
| Chemical Name or Material | Norethindrone |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction