Learn More
GPE Scientific™ Norell™ Coaxial Inserts For NMR Sample Tubes
Product Code. 12812316
Change view
Click to view available options
Capacity (Metric):
100μL
120μL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12812316 100μL Each
12822316 120μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12812316 Supplier GPE Scientific™ Supplier No. NI5CCIB
Only null left
Add to basket
Add to basket
Specifications
Includes Cap for the insert tube and special cap to seal the insert to the top of the outer NMR tube
Product Type Coaxial Insert
Length (Metric) 206 mm
Capacity (Metric) 100μL