missing translation for 'onlineSavingsMsg'
Learn More

Norell™ Borosilicate Glass J Young NMR Valves

Brand:  Norell S-5-600-JY-8-YELLOW

Code : 50

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12802316

  • £132.00 / Each


Estimated Shipment
04-04-2018
Add to basket

Specifications

Specifications

J Young NMR Valve
203mm
Female joint
Borosilicate Glass
600MHz tube
1Each
Videos
Documentation