missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NORE1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB500-248SS
This item is not returnable.
View return policy
Description
NORE1 Polyclonal specifically detects NORE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| NORE1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Maxp1, MGC10823, MGC17344, New ras effector 1, NORE1A, NORE1B, NORE1RASSF3, Rap1-binding protein, RAPLnovel Ras effector 1, Ras association (RalGDS/AF-6) domain family 5, Ras association (RalGDS/AF-6) domain family member 5, ras association domain-containing protein 5, Ras effector-like protein, regulator for cell adhesion and polarization enriched in lymphoid tissue, Regulator for cell adhesion and polarization enriched in lymphoid tissues, tumor suppressor RASSF3 | |
| Rabbit | |
| 47.1 kDa | |
| 0.025 mL | |
| Cancer, Signal Transduction, Tumor Suppressors | |
| 83593 | |
| Mouse, Human, Rat | |
| IgG |
| Immunohistochemistry, Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:500 -1:1000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:150, Immunohistochemistry-Paraffin 1:150 | |
| Q8WWW0 | |
| RASSF5 | |
| Residues 313-329 [DNPQKFALFKRIHKDGQ] of the NORE1 protein. This sequence is located in the RA domain of NORE1. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This is specific for NORE1, accession number NP_113625.1. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction