missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nordoxepin.HCl, 1mg/ml in Methanol (as free base), Lipomed
Description
Nordoxepin.HCl, 1mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Nordoxepin Hydrochloride |
| CAS | 2887-91-4 |
| Analyte or Component Names | Nordoxepin Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H19 N O . HCl |
| Concentration | 1 mg/mL |
| Synonym | 1-Propanamine, 3-dibenz[b,e]oxepin-11(6 H)-ylidene-N-methyl-, hydrochloride (1:1), 1-Propanamine, 3-dibenz[b,e]oxepin-11(6 H)-ylidene-N-methyl-, hydrochloride (9 CI), Dibenz[b,e]oxepin-Δ11(6 H),γ-propylamine, N-methyl-, hydrochloride (7 CI,8 CI), Dibenz[b,e]oxepin, 1-propanamine deriv., Desmethyldoxepine hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6 H)-ylidene)-N-methylpropan-1-amine hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6 H)-ylidene)-N-methylpropan-1-amine hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6 H)-ylidene)-N-methylpropan-1-amine hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6 H)-ylidene)-N-methylpropan-1-amine hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6 H)-ylidene)-N-methylpropan-1-amine hydrochloride, (EZ)-3-(Dibenzo[b,e]oxepin-11(6 H)-ylidene)-N-methylpropan-1-amine hydrochloride |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CNCCC=C1c2ccccc2COc3ccccc13 |
| IUPAC Name | 3-(6 H-benzo[c][1 ]benzoxepin-11-ylidene)-N-methylpropan-1-amine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?