missing translation for 'onlineSavingsMsg'
Learn More

Nordihydroguaiaretic acid, MedChemExpress

Product Code. 16409036
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16409036 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16409036 Supplier Medchem Express Supplier No. HYN0198
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nordihydroguaiaretic acid is a 5-lipoxygenase (5LOX) (IC50=8 μM) and tyrosine kinase inhibitor.

Nordihydroguaiaretic acid is a 5-lipoxygenase (5LOX) (IC50=8 μM) and tyrosine kinase inhibitor.

Specifications

Chemical Name or Material Nordihydroguaiaretic acid
CAS 500-38-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H22O4
Quantity 100 mg
Synonym NDGA
Solubility Information DMSO : 250 mg/mL (826.83 mM; Need ultrasonic)
SMILES CC(CC1=CC=C(O)C(O)=C1)C(C)CC2=CC=C(O)C(O)=C2
Molecular Weight (g/mol) 302.36
Formula Weight 302.36
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.