missing translation for 'onlineSavingsMsg'
Learn More

Nordihydroguaiaretic acid, 97%

Product Code. p-7043862
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11372036

Brand: Thermo Scientific Alfa Aesar L03149.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 500-38-9 | C18H22O4 | 302.37 g/mol

Nordihydroguaiaretic acid is used in the treatment of multiple diseases, including cardiovascular diseases, neurological disorders and cancers. It is also used as an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in 100% ethanol, methanol, acetone, dimethylsulfoxide, dimethylformamide, ethanol (5% w/v), glycol, PEG, and slightly soluble in hot water. Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500-38-9
Molecular Formula C18H22O4
Molecular Weight (g/mol) 302.37
MDL Number MFCD00002206
InChI Key HCZKYJDFEPMADG-UHFFFAOYSA-N
Synonym nordihydroguaiaretic acid, ndga, dihydronorguaiaretic acid, norhydroguaiaretic acid, nordihydroguairaretic acid, norguaiaretic acid, dihydro, 1,2-benzenediol, 4,4'-2,3-dimethyl-1,4-butanediyl bis, dinorguaiaretic acid, dihydro, 4,4'-2,3-dimethyltetramethylene dipyrocatechol, 2,3-bis 3,4-dihydroxyphenylmethyl butane
PubChem CID 4534
ChEBI CHEBI:7625
IUPAC Name 4-[4-(3,4-dihydroxyphenyl)-2,3-dimethylbutyl]benzene-1,2-diol
SMILES CC(CC1=CC(=C(C=C1)O)O)C(C)CC2=CC(=C(C=C2)O)O

Specifications

Melting Point 184°C to 188°C
Odor Odorless
Quantity 1 g
Beilstein 2056825
Merck Index 14,6693
Solubility Information Soluble in 100% ethanol,methanol,acetone,dimethylsulfoxide,dimethylformamide,ethanol (5% w/v),glycol,PEG,and slightly soluble in hot water. Insoluble in water.
Formula Weight 302.37
Percent Purity 97%
Chemical Name or Material Nordihydroguaiaretic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.