missing translation for 'onlineSavingsMsg'
Learn More

Nordihydro Guaiaretic Acid (mixture of diastereomers), TRC

Product Code. 30179872
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179872 250 mg 250mg
30179811 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179872 Supplier Toronto Research Chemicals Supplier No. TRCN673000250MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500-38-9
Molecular Formula C18 H22 O4
Molecular Weight (g/mol) 302.36
Synonym 1,2-Benzenediol, 4,4'-(2,3-dimethyl-1,4-butanediyl)bis-, Pyrocatechol, 4,4'-(2,3-dimethyltetramethylene)di- (8CI), 1,4-Bis(3,4-dihydroxyphenyl)-2,3-dimethylbutane, 4,4'-(2,3-Dimethyl-1,4-butanediyl)bis(pyrocatechol), 4,4'-(2,3-Dimethyl-1,4-butanediyl)bis[1,2-benzenediol], 4,4'-(2,3-Dimethyltetramethylene)dipyrocatechol, 4-[4-(3,4-Dihydroxyphenyl)-2,3-dimethylbutyl]benzene-1,2-diol, Butane, 1,4-bis(3,4-dihydroxyphenyl)-2,3-dimethyl-, Dihydronorguaiaretic acid, Dinorguaiaretic acid, dihydro-, NDGA, NSC 4291, Nordihydroguaiaretic acid, Norguaiaretic acid, dihydro-, β,γ-Dimethyl-α,δ-bis(3,4-dihydroxyphenyl)butane
IUPAC Name 4-[4-(3,4-dihydroxyphenyl)-2,3-dimethylbutyl]benzene-1,2-diol
SMILES CC(Cc1ccc(O)c(O)c1)C(C)Cc2ccc(O)c(O)c2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C18H22O4/c1-11(7-13-3-5-15(19)17(21)9-13)12(2)8-14-4-6-16(20)18(22)10-14/h3-6,9-12,19-22H,7-8H2,1-2H3
Formula Weight 302.1518
Chemical Name or Material Nordihydroguaiaretic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.