missing translation for 'onlineSavingsMsg'
Learn More

Norchlorprothixene, TRC

Product Code. 30413017
Change view
Click to view available options
Quantity:
10 mg
100 mg
Product Code. Quantity
30413017 100 mg
30420819 10 mg
2 options
This item is not returnable. View return policy

Product Code. 30413017

Brand: Toronto Research Chemicals TRCN662100100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51382-91-3
Formula Weight 301.0692
InChI Formula InChI=1S/C17H16ClNS/c1-19-10-4-6-13-14-5-2-3-7-16(14)20-17-9-8-12(18)11-15(13)17/h2-3,5-9,11,19H,4,10H2,1H3/b13-6-
IUPAC Name (3Z)-3-(2-chlorothioxanthen-9-ylidene)-N-methylpropan-1-amine
Molecular Formula C17H16ClNS
Molecular Weight (g/mol) 301.83
SMILES CNCC\C=C/1\c2ccccc2Sc3ccc(Cl)cc13
Synonym 9H-Thioxanthene, 1-Propanamine Deriv., Demethylchlorprothixene, N-Monodemethylchlorprothixene, (Z)-3-(2-Chloro-9H-thioxanthen-9-ylidene)-N-methyl-1-propanamine, (3Z)-3-(2-Chloro-9H-thioxanthen-9-ylidene)-N-methyl-1-propanamine
Chemical Name or Material Norchlorprothixene

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.