missing translation for 'onlineSavingsMsg'
Learn More

Norcamphor, 98+%

Product Code. 11351297
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11351297 5 g 5g
11361297 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351297 Supplier Thermo Scientific Alfa Aesar Supplier No. L01491.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a building block in organic synthesis and also acts as a precursor to norborneols. | CAS: 497-38-1 | C7H10O | 110.156 g/mol

Norcamphor is used as a building block in organic synthesis. It is also used as a precursor to norborneols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Norcamphor is used as a building block in organic synthesis. It is also used as a precursor to norborneols.

Solubility
Insoluble in water. Soluble in methanol.

Notes
Incompatible with strong oxidizing agents. Heat and light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-38-1
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.156
MDL Number MFCD00074823
InChI Key KPMKEVXVVHNIEY-UHFFFAOYSA-N
Synonym norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one
PubChem CID 10345
IUPAC Name bicyclo[2.2.1]heptan-3-one
SMILES C1CC2CC1CC2=O

Specifications

Melting Point 92°C to 97°C
Boiling Point 172°C to 174°C
Flash Point 33°C (91°F)
Quantity 5 g
UN Number UN1325
Beilstein 1209657
Solubility Information Insoluble in water. Soluble in methanol.
Formula Weight 110.16
Percent Purity ≥98%
Chemical Name or Material Norcamphor

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.