missing translation for 'onlineSavingsMsg'
Learn More

(Norbornadiene)rhodium(I) 2,4-pentanedionate, Rh 34.9%, Thermo Scientific™

Product Code. 15457295
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
missing translation for 'unitSize'
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457295 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15457295 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' 046727.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 32354-50-0
Melting Point 176°C to 178°C
Color Yellow
Molecular Formula C12H15O2Rh
MDL Number MFCD00075071
Quantity 1 g
Synonym Acetylacetonato(norbornadiene)rhodium(I); Ir(acac)(nbd)
InChI Key IIQPRGKBFZGPIB-LWFKIUJUSA-M
SMILES [Rh+].C\C([O-])=C\C(C)=O.C1C2C=CC1C=C2
IUPAC Name λ¹-rhodium(1+) bicyclo[2.2.1]hepta-2,5-diene (2Z)-4-oxopent-2-en-2-olate
Molecular Weight (g/mol) 294.16
Formula Weight 294.15
Concentration or Composition (by Analyte or Components) Rh 34.9%
Physical Form Crystalline Solid
Chemical Name or Material (Norbornadiene)rhodium(I) 2,4-pentanedionate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.