missing translation for 'onlineSavingsMsg'
Learn More

Nor Verapamil Hydrochloride, TRC

Product Code. 18093130
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18093130 5 mg 5mg
18042430 50 mg 50mg
18014030 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18093130 Supplier Toronto Research Chemicals Supplier No. TRCN8775005MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67812-42-4
Molecular Formula C26H37ClN2O4
Molecular Weight (g/mol) 477.04
Synonym Benzeneacetonitrile, α-[3-[[2-(3,4-dimethoxyphenyl)ethyl]amino]propyl]-3,4-dimethoxy-α-(1-methylethyl)-, hydrochloride (1:1), Benzeneacetonitrile, α-[3-[[2-(3,4-dimethoxyphenyl)ethyl]amino]propyl]-3,4-dimethoxy-α-(1-methylethyl)-, monohydrochloride (9CI), Valeronitrile, 5-[(3,4-dimethoxyphenethyl)amino]-2-(3,4-dimethoxyphenyl)-2-isopropyl-, hydrochloride (7CI), D 591 HCl, Norverapamil hydrochloride, (2RS)-2-(3,4-Dimethoxyphenyl)-5-[[2-(3,4-dimethoxyphenyl)-ethyl]amino]-2-(1-methylethyl)pentanenitrile hydrochloride, N-Norverapamil hydrochloride
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethylamino]-2-propan-2-ylpentanenitrile;hydrochloride
SMILES Cl.COc1ccc(CCNCCCC(C#N)(C(C)C)c2ccc(OC)c(OC)c2)cc1OC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C26H36N2O4.ClH/c1-19(2)26(18-27,21-9-11-23(30-4)25(17-21)32-6)13-7-14-28-15-12-20-8-10-22(29-3)24(16-20)31-5;/h8-11,16-17,19,28H,7,12-15H2,1-6H3;1H
Formula Weight 476.2442
Percent Purity >95%
Chemical Name or Material Norverapamil Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.