missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Nor NOHA monoacetate
GREENER_CHOICE

Product Code. 18748248
Change view
Click to view available options
:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18748248 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18748248 Supplier Tocris Bioscience™ Supplier No. 6370/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Arginase inhibitor

Nor NOHA monoacetate is a reversible, competitive arginase inhibitor (IC50 = 2 μM). Exhibits 10-fold selectivity for human type II arginase over type I. Enhances the effect of acetylcholine on isolated aortic and mesenteric arterial rings. Inhibits growth of lung carcinoma implants in mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S)-2-Amino-4-[[(hydroxyamino)iminomethyl]amino]butanoic acid monoacetate
CAS 2250019-93-1
Quantity 10 mg
Molecular Formula C5H12N4O3.C2H4O2
Formula Weight Observed MW: 236.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.