missing translation for 'onlineSavingsMsg'
Learn More

nor-NOHA acetate, MedChemExpress

Product Code. 16497437
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16497437

Brand: Medchem Express HY112885A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress nor-NOHA acetate (Nω-Hydroxy-nor-L-arginine acetate) is a specific and reversible arginase inhibitor, induces apoptosis in ARG2-expressing cells under hypoxia but not normoxia. Anti-leukemic activity, effective in endothelial dysfunction, immunosuppression and metabolism.

nor-NOHA acetate (Nω-Hydroxy-nor-L-arginine acetate) is a specific and reversible arginase inhibitor, induces apoptosis in ARG2-expressing cells under hypoxia but not normoxia. Anti-leukemic activity, effective in endothelial dysfunction, immunosuppression and metabolism.

Specifications

Chemical Name or Material nor-NOHA acetate
Physical Form Solid
CAS 1140844-63-8
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C9H20N4O7
For Use With (Application) Cancer-programmed cell death
Synonym Nω-Hydroxy-nor-L-arginine acetate
Solubility Information H2O : 200 mg/mL (675.04 mM; Need ultrasonic)
SMILES O=C(O)[C@@H](N)CCNC(NO)=N.CC(O)=O.CC(O)=O
Molecular Weight (g/mol) 296.28
Formula Weight 296.28
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.