missing translation for 'onlineSavingsMsg'
Learn More

Nor Mianserin, TRC

Product Code. 30413018
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Product Code. Quantity
30413018 5 mg
30420354 10 mg
30428712 50 mg
3 options
This item is not returnable. View return policy

Product Code. 30413018

Brand: Toronto Research Chemicals TRCN7368005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71936-92-0
Formula Weight 250.147
InChI Formula InChI=1S/C17H18N2/c1-3-7-15-13(5-1)11-14-6-2-4-8-16(14)19-10-9-18-12-17(15)19/h1-8,17-18H,9-12H2
Molecular Formula C17 H18 N2
Molecular Weight (g/mol) 250.34
SMILES C1CN2C(CN1)c3ccccc3Cc4ccccc24
Synonym Dibenzo[c,f]pyrazino[1,2-a]azepine, 1,2,3,4,10,14b-hexahydro-, 1,2,3,4,10,14b-Hexahydrodibenzo[c,f]pyrazino[1,2-a]azepine, (±)-Demethylmianserin, Demethylmianserin, Desmethylmianserin, N-Desmethylmianserin, Normianserin, Normianserine, Mianserin Hydrochloride Imp. E (EP)
Chemical Name or Material Nor Mianserin

Specifications

Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.